Merck
  • Vascular endothelial growth factor up-regulates expression of receptor activator of NF-kappa B (RANK) in endothelial cells. Concomitant increase of angiogenic responses to RANK ligand.
Vascular endothelial growth factor up-regulates expression of receptor activator of NF-kappa B (RANK) in endothelial cells. Concomitant increase of angiogenic responses to RANK ligand.
The Journal of biological chemistry (2003-08-02)
Jeong-Ki Min, Young-Myeong Kim, Young-Mi Kim, Eok-Cheon Kim, Yong Song Gho, Il-Jun Kang, Soo-Young Lee, Young-Yun Kong, Young-Guen Kwon
ABSTRACT
Vascular endothelial growth factor (VEGF) is known as a key regulator of angiogenesis during endochondral bone formation. Recently, we demonstrated that TNF-related activation-induced cytokine (TRANCE or RANKL), which is essential for bone remodeling, also had an angiogenic activity. Here we report that VEGF up-regulates expression of receptor activator of NF-kappa B (RANK) and increases angiogenic responses of endothelial cells to TRANCE. Treatment of human umbilical vein endothelial cells (HUVECs) with VEGF increased both RANK mRNA and surface protein expression. Although placenta growth factor specific to VEGF receptor-1 had no significant effect on RANK expression, inhibition of downstream signaling molecules of the VEGF receptor-2 (Flk-1/KDR) such as Src, phospholipase C, protein kinase C, and phosphatidylinositol 3'-kinase suppressed VEGF-stimulated RANK expression in HUVECs. Moreover, the MEK inhibitor PD98059 or expression of dominant negative MEK1 inhibited induction of RANK by VEGF but not the Ca2+ chelator 1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid-acetoxymethyl ester (BAPTA-AM). VEGF potentiated TRANCE-induced ERK activation and tube formation via RANK up-regulation in HUVECs. Together, these results show that VEGF enhances RANK expression in endothelial cells through Flk-1/KDR-protein kinase C-ERK signaling pathway, suggesting that VEGF plays an important role in modulating the angiogenic action of TRANCE under physiological or pathological conditions.