Merck
  • Separation and purification of baicalin and wogonoside from the Chinese medicinal plant Scutellaria baicalensis Georgi by high-speed counter-current chromatography.
Separation and purification of baicalin and wogonoside from the Chinese medicinal plant Scutellaria baicalensis Georgi by high-speed counter-current chromatography.
Journal of chromatography. A (2005-03-30)
Sujuan Wu, Ailing Sun, Renmin Liu
ABSTRACT
A preparative high-speed counter-current chromatography (HSCCC) method for isolation and purification of baicalin and wogonoside from the Chinese medicinal plant Scutellaria baicalensis Georgi (Huang-qin in Chinese) was successfully established by using ethyl acetate-methanol-1% acetic acid water (5:0.5:5, v/v) as the two-phase solvent system. The upper phase of ethyl acetate-methanol-1% acetic acid water (5:0.5:5, v/v) was used as the stationary phase of HSCCC. Baicalin (58.1 mg) and wogonoside (17.0mg) with the purity of 99.2 and 99.0%, respectively, were separated successfully in one-step separation from 120 mg of crude sample from S. baicalensi, Georgi. The structures of baicalin and wogonoside were identified by 1H NMR and 13C NMR.
Sigma-Aldrich
Wogonoside, ≥95% (LC/MS-ELSD)