Merck
  • Effects of the H(3) antagonist, thioperamide, on behavioral alterations induced by systemic MK-801 administration in rats.
Effects of the H(3) antagonist, thioperamide, on behavioral alterations induced by systemic MK-801 administration in rats.
Psychopharmacology (2009-05-26)
Mark E Bardgett, Megan Points, John Roflow, Meredith Blankenship, Molly S Griffith
ABSTRACT
Recent studies have raised the possibility that antagonists of H(3) histamine receptors possess cognitive-enhancing and antipsychotic properties. However, little work has assessed these compounds in classic animal models of schizophrenia. The purpose of this study was to determine if a prototypical H(3) antagonist, thioperamide, could alter behavioral deficits caused by the N-methyl-D: -aspartate (NMDA) receptor antagonist, MK-801, in adult male rats. MK-801 was chosen to be studied since it produces a state of NMDA receptor hypofunction in rats that may be analogous to the one hypothesized to occur in schizophrenia. The interaction between thioperamide and MK-801 was measured in three behavioral tests: locomotor activity, prepulse inhibition (PPI), and delayed spatial alternation. In each test, rats received a subcutaneous injection of saline or thioperamide (3.0 and 10 mg/kg) followed 20 min later by a subcutaneous injection of saline or MK-801 (0.05, 0.10, and 0.30 mg/kg). Locomotor activity was significantly elevated by MK-801 in a dose-dependent manner. Thioperamide pretreatment alone did not alter locomotor activity; however, its impact on MK-801 was dose-dependent. Each thioperamide dose enhanced the effects of two lower doses of MK-801 but reduced the effect of a higher MK-801 dose. Clear deficits in PPI and delayed spatial alternation were produced by MK-801 treatment, but neither impairment was significantly modified by thioperamide pretreatment. H(3) receptors modulate responses to NMDA antagonists in behaviorally specific and dose-dependent ways.