Merck
  • Messenger RNA and protein expression of thymidylate synthase and DNA repair genes in thymic tumors.
Messenger RNA and protein expression of thymidylate synthase and DNA repair genes in thymic tumors.
Lung cancer (Amsterdam, Netherlands) (2013-01-02)
Valentina Monica, Ubaldo Familiari, Luigi Chiusa, Giulio Rossi, Domenico Novero, Simone Busso, Enrico Ruffini, Francesco Ardissone, Giorgio V Scagliotti, Mauro Papotti
ABSTRACT
Thymic epithelial tumors include several entities with different biologic behavior. Chemotherapy is indicated in advanced disease, but limited data exist on gene expression correlation with the response to chemotherapeutic agents. A series of 69 thymic neoplasms (7 A-, 6 AB-, 6 B1-, 10 B2-, 14 B3-thymomas, 22 carcinomas and 4 combined tumors) was collected to assess gene expression of thymidylate synthase (TS), excision repair cross complementing-1 (ERCC1), ribonucleotide reductase subunit 1 (RRM1), topoisomerase 2α (TOP2A) and mTOR. A strong linear correlation between TS gene and protein expression was observed (P<0.0001, R=0.40). TS expression was significantly lower in pure A-thymomas and thymic carcinomas (P<0.0001) and progressively decreasing from B1-type to thymic carcinomas (B1>B2>B3>C; P<0.0001). RRM1 and TOP2A mRNA expression levels were significantly correlated with TS levels (both P=0.03) with a similar trend of expression among histotypes. RRM1 and TOP2A high levels were significantly correlated with high TS (P=0.03) and low tumor stages (I-II) (P<0.0001 and P<0.01, respectively). No relevant changes of ERCC1 and mTOR were detected. Low TS and, to a minor extent, RRM1 and TOP2A expression were detected in aggressive thymic tumors. These findings should be prospectively considered in selecting the most appropriate chemotherapy.
Sigma-Aldrich
Topoisomerase II α human, (Single band on SDS-PAGE), liquid
Sigma-Aldrich
DNA Gyrase from Escherichia coli, aqueous glycerol solution