Merck
Cloning and expression of murine immune interferon cDNA.
Proceedings of the National Academy of Sciences of the United States of America (1983-10-01)
P W Gray, D V Goeddel
ABSTRACT
The murine immune interferon (IFN-gamma) gene was cloned and expressed under control of the simian virus 40 early promoter in the monkey COS-1 cell line. A protein is secreted from these cells having the biological, antigenic, and biochemical characteristics of natural murine IFN-gamma. Cloned murine IFN-gamma cDNAs were obtained by using RNA from both mitogen-induced murine spleens and the transfected COS cells, and both code for identical proteins. The mature murine IFN-gamma encoded is 136 amino acids long, 10 amino acids shorter than human IFN-gamma. The nucleotide homology between the murine and human IFN-gamma genes is 60-65%, whereas the encoded proteins are only 40% homologous. Murine IFN-gamma cDNA was expressed in Escherichia coli under trp promoter control.
Sigma-Aldrich
Interferon-γ from mouse, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture