Merck
  • Somatic cell hybrids producing antibodies specific to human fibronectin.
Somatic cell hybrids producing antibodies specific to human fibronectin.
International journal of cancer (1980-03-15)
L Zardi, B Carnemolla, A Siri, L Santi, R S Accolla
ABSTRACT
We have produced somatic cell hybrids between mouse plasocytoma cells deficient in hypoxanthine phosphoribosyltransferase (P3 x 63 Ag8) and spleen cells derived from mice immunized with purified human plasma fibronectin. We report herein the properties of monoclonal anti-fibronectin antibodies released by eight different clones.
Sigma-Aldrich
Anti-Fibronectin antibody, Mouse monoclonal, clone IST-3, purified from hybridoma cell culture