MilliporeSigma
  • DR6 Augments Colorectal Cancer Cell Growth, Invasion, and Stemness by Activating AKT/NF-κB Pathway.
DR6 Augments Colorectal Cancer Cell Growth, Invasion, and Stemness by Activating AKT/NF-κB Pathway.
Biochemical genetics (2024-03-13)
Jing Jia, Yisen Huang, Qiwei Chen, Jianbin Hou, Yan Liu, Lifeng Xie, Xinyu Li, Chunkang Yang
ABSTRACT
This study aims to elucidate the role and mechanisms of Death Receptor 6 (DR6), a member of the tumor necrosis factor receptor superfamily, in the malignant progression of colorectal cancer (CRC). The association of DR6 expression levels and CRC patient survival was examined using the CRC cohort data from GEPIA database. The functional role of DR6 in CRC cells was investigated by performing loss-of-function and gain-of-function experiments based on CCK-8 proliferation assay, transwell migration and invasion assay, and sphere-forming assays. Xenograft model of CRC cells in nude mouse was established to evaluate the impact of DR6 knockdown on CRC tumorigenesis. Elevated expression of DR6 was correlated with an unfavorable prognosis in CRC patients. In vitro functional assays demonstrated that silencing DR6 considerably suppressed the proliferation, migration, invasion, and stemness of CRC cells, whereas its overexpression showed an opposite effect. DR6 knockdown also attenuated tumor formation of CRC cells in the nude mice. Mechanistically, silencing DR6 reduced the phosphorylation of AKT and NF-κB in CRC cells, and the treatment with an AKT activator (SC79) abrogated the inhibitory effects of DR6 knockdown on the malignant features of CRC cells. Our data suggest that DR6 contributes to the malignant progression of CRC by activating AKT/NF-κB pathway, indicating its clinical potential as a prognostic marker and therapeutic target for CRC.
Sigma-Aldrich
Anti-TNFRSF21 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution