Merck
  • Glycerol kinase enzyme is a prognostic predictor in esophageal carcinoma and is associated with immune cell infiltration.
Glycerol kinase enzyme is a prognostic predictor in esophageal carcinoma and is associated with immune cell infiltration.
Scientific reports (2024-02-17)
Fei Ying, Xuyong Chen, Lihong Lv
ABSTRACT
The influence of lipid metabolism on tumorigenesis and progression has garnered significant attention. However, the role of Glycerol Kinase (GK), a key enzyme in glycerol metabolism, in Esophageal Carcinoma (ESCA) remains unclear. To further elucidate the relationship between GK and ESCA, we investigated GK expression levels using database information. Controlled studies employing immunohistochemistry were conducted on clinical ESCA tumor samples and normal specimens, confirming GK's elevated expression in ESCA. Analysis of The Cancer Genome Atlas (TCGA) data via Kaplan-Meier (KM) survival plots revealed that increased GK expression correlates with poorer ESCA patient outcomes, particularly in overall survival (OS) and disease-specific survival (DSS). Multiple regression analysis indicated that elevated GK expression is an independent risk factor affecting ESCA prognosis. Statistical analysis of prognostic data from clinical samples further corroborated this finding. Moreover, there appears to be a significant correlation between GK expression and immune infiltration, specifically involving certain T and B lymphocytes. In conclusion, elevated GK expression in ESCA is strongly linked to poor prognosis and increased immune cell infiltration, highlighting its potential as an independent prognostic biomarker and a viable therapeutic target.
Sigma-Aldrich
Glycerokinase from Cellulomonas sp., lyophilized powder, 25-75 units/mg protein