Merck
  • Generation of B-cell-based cellular vaccine for cancer in murine models.
Generation of B-cell-based cellular vaccine for cancer in murine models.
STAR protocols (2023-04-21)
David Hou, Joshua L Katz, Catalina Lee-Chang
ABSTRACT
A B-cell-based cellular vaccine (BVax), produced from 4-1BBL+ B cells, can select tumor-specific B cells that, upon ex vivo culture, can generate tumor-specific antibodies and activate T cells. Here, we present a protocol to generate a B-cell-based vaccine in a CT2A orthotopic glioma murine model. We describe steps for BVax production involving glioma cell implantation, tissue harvesting, 4-1BBL+ B cell isolation, and activation. We also describe assessing BVax phenotype in vitro and in vivo functional status. For complete details on the use and execution of this protocol, please refer to Lee-Chang et al. (2021).1.
Sigma-Aldrich
CT-2A Mouse Glioma Cell Line, CT-2A mouse glioma cell line is a valuable mouse model for therapeutic research on brain malignancies.