Merck
  • Single-cell multiomic analysis identifies a HOX-PBX gene network regulating the survival of lymphangioleiomyomatosis cells.
Single-cell multiomic analysis identifies a HOX-PBX gene network regulating the survival of lymphangioleiomyomatosis cells.
Science advances (2023-05-10)
Tasnim Olatoke, Andrew Wagner, Aristotelis Astrinidis, Erik Y Zhang, Minzhe Guo, Alan G Zhang, Ushodaya Mattam, Elizabeth J Kopras, Nishant Gupta, Eric P Smith, Magdalena Karbowniczek, Maciej M Markiewski, Kathryn A Wikenheiser-Brokamp, Jeffrey A Whitsett, Francis X McCormack, Yan Xu, Jane J Yu
ABSTRACT
Lymphangioleiomyomatosis (LAM) is a rare, progressive lung disease that predominantly affects women. LAM cells carry TSC1/TSC2 mutations, causing mTORC1 hyperactivation and uncontrolled cell growth. mTORC1 inhibitors stabilize lung function; however, sustained efficacy requires long-term administration, and some patients fail to tolerate or respond to therapy. Although the genetic basis of LAM is known, mechanisms underlying LAM pathogenesis remain elusive. We integrated single-cell RNA sequencing and single-nuclei ATAC-seq of LAM lungs to construct a gene regulatory network controlling the transcriptional program of LAM cells. We identified activation of uterine-specific HOX-PBX transcriptional programs in pulmonary LAMCORE cells as regulators of cell survival depending upon HOXD11-PBX1 dimerization. Accordingly, blockage of HOXD11-PBX1 dimerization by HXR9 suppressed LAM cell survival in vitro and in vivo. PBX1 regulated STAT1/3, increased the expression of antiapoptotic genes, and promoted LAM cell survival in vitro. The HOX-PBX gene network provides promising targets for treatment of LAM/TSC mTORC1-hyperactive cancers.
Sigma-Aldrich
Monoclonal Anti-HOXD11, (N-terminal) antibody produced in mouse, clone 6C8, purified immunoglobulin, buffered aqueous solution