Merck
  • Orexin recruits non-selective cationic conductance and endocannabinoid to dynamically modulate firing of caudal pontine reticular nuclear neurones.
Orexin recruits non-selective cationic conductance and endocannabinoid to dynamically modulate firing of caudal pontine reticular nuclear neurones.
The Journal of physiology (2023-07-08)
Qi-Cheng Qiao, Si-Yi Wen, Yi-Bin Jiang, Hui Feng, Rui Xu, Yan-Jia Huang, Bang-Yun Chen, Wen-Hao Chen, Jia-Hui Niu, Rong Hu, Nian Yang, Jun Zhang
ABSTRACT
The neuropeptide orexin is involved in motor circuit function. However, its modulation on neuronal activities of motor structures, integrating orexin's diverse downstream molecular cascades, remains elusive. By combining whole-cell patch-clamp recordings and neuropharmacological methods, we revealed that both non-selective cationic conductance (NSCC) and endocannabinoids (eCBs) are recruited by orexin signalling on reticulospinal neurones in the caudal pontine reticular nucleus (PnC). The orexin-NSCC cascade provides a depolarizing force that proportionally enhances the firing-responsive gain of these neurones. Meanwhile, the orexin-eCB cascade selectively attenuates excitatory synaptic strength in these neurones by activating presynaptic cannabinoid receptor type 1. This cascade restrains the firing response of the PnC reticulospinal neurones to excitatory inputs. Intriguingly, non-linear or linear interactions between orexin postsynaptic excitation and presynaptic inhibition can influence the firing responses of PnC reticulospinal neurones in different directions. When presynaptic inhibition is in the lead, non-linear interactions can prominently downregulate or even gate the firing response. Conversely, linear interactions occur to promote the firing response, and these linear interactions can be considered a proportional reduction in the contribution of depolarization to firing by presynaptic inhibition. Through the dynamic employment of these interactions, adaptive modulation may be achieved by orexin to restrain or even gate the firing output of the PnC to weak/irrelevant input signals and facilitate those to salient signals. KEY POINTS: This study investigated the effects of orexin on the firing activity of PnC reticulospinal neurones, a key element of central motor control. We found that orexin recruited both the non-selective cationic conductances (NSCCs) and endocannabinoid (eCB)-cannabinoid receptor type 1 (CB1R) system to pontine reticular nucleus (PnC) reticulospinal neurones. The orexin-NSCC cascade exerts a postsynaptic excitation that enhances the firing response, whereas the orexin-eCB-CB1R cascade selectively attenuates excitatory synaptic strength that restrains the firing response. The postsynaptic and presynaptic actions of orexins occur in an overlapping time window and interact to dynamically modulate firings in PnC reticulospinal neurones. Non-linear interactions occur when presynaptic inhibition of orexin is in the lead, and these interactions can prominently downregulate or even gate firing responses in PnC reticulospinal neurones. Linear interactions occur when postsynaptic excitation of orexin is in the lead, and these interactions can promote the firing response. These linear interactions can be considered a proportional reduction of the contribution of depolarization to firing by presynaptic inhibition.
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-Glutamate antibody produced in mouse, clone GLU-4, ascites fluid
Sigma-Aldrich
Anti-Cannabinoid Receptor 1 Antibody, Chemicon®, from rabbit