Merck
  • Protocol to analyze the transmigration efficiency of T. brucei using an in vitro model of the blood-cerebrospinal fluid barrier.
Protocol to analyze the transmigration efficiency of T. brucei using an in vitro model of the blood-cerebrospinal fluid barrier.
STAR protocols (2022-09-15)
Alexander Herrmann, Carolin Susanne Schnedermann, Hiroshi Ishikawa, Christian Schwerk, Horst Schroten, Stefan Mogk
ABSTRACT
At present, the only approach to investigate the transmigration of Trypanosoma brucei, the causative agent of human African trypanosomiasis, from blood to cerebrospinal fluid is through animal experiments. This protocol details how to analyze the transmigration efficiency using an in vitro model of the blood-cerebrospinal fluid (blood-CSF) barrier. We describe how to grow human choroid plexus epithelial cells on cell culture filter inserts to form the barrier, followed by isolating and quantifying genomic DNA of transmigrated parasites by qPCR. For complete details on the use and execution of this protocol, please refer to Speidel et al. (2022).
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture