Merck
  • Meldonium improves Huntington's disease mitochondrial dysfunction by restoring peroxisome proliferator-activated receptor γ coactivator 1α expression.
Meldonium improves Huntington's disease mitochondrial dysfunction by restoring peroxisome proliferator-activated receptor γ coactivator 1α expression.
Journal of cellular physiology (2018-10-27)
Francesca Di Cristo, Mauro Finicelli, Filomena Anna Digilio, Simona Paladino, Anna Valentino, Filippo Scialò, Maria D'Apolito, Carmela Saturnino, Umberto Galderisi, Antonio Giordano, Mariarosa Anna Beatrice Melone, Gianfranco Peluso
ABSTRACT
Mitochondrial dysfunction seems to play a fundamental role in the pathogenesis of neurodegeneration in Huntington's disease (HD). We assessed possible neuroprotective actions of meldonium, a small molecule affecting mitochondrial fuel metabolism, in in vitro and in vivo HD models. We found that meldonium was able to prevent cytotoxicity induced by serum deprivation, to reduce the accumulation of mutated huntingtin (mHtt) aggregates, and to upregulate the expression of peroxisome proliferator-activated receptor γ coactivator 1α (PGC-1α) in mHTT-expressing cells. The PGC-1α increase was accompanied by the increment of mitochondrial mass and by the rebalancing of mitochondrial dynamics with a promotion of the mitochondrial fusion. Meldonium-induced PGC-1α significantly alleviated motor dysfunction and prolonged the survival of a transgenic HD Drosophila model in which mHtt expression in the nervous system led to progressive motor performance deficits. Our study strongly suggests that PGC-1α, as a master coregulator of mitochondrial biogenesis, energy homeostasis, and antioxidant defense, is a potential therapeutic target in HD.
Sigma-Aldrich
Anti-GAPDH Antibody, from chicken, purified by affinity chromatography
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Polyglutamine-Expansion Diseases Marker Antibody, clone 5TF1-1C2, ascites fluid, clone 5TF1-1C2, Chemicon®