Merck
  • Flow Cytometry Protocols for Assessment of Platelet Function in Whole Blood.
Flow Cytometry Protocols for Assessment of Platelet Function in Whole Blood.
Methods in molecular biology (Clifton, N.J.) (2017-08-15)
Leonardo Pasalic, Gabrielle J Pennings, David Connor, Heather Campbell, Leonard Kritharides, Vivien M Chen
ABSTRACT
Flow cytometry is a powerful tool for rapid evaluation of multiple functional properties of large numbers of platelets in whole blood. In the following chapter, we provide a number of flow cytometry-based protocols broadly aimed at (1) assessment of constitutively expressed platelet membrane receptors to diagnose inherited platelet bleeding disorders and (2) investigation of basal and agonist-induced platelet functional responses including generation of platelet-leukocyte aggregates, alpha and dense granule release, calcium flux, and phosphatidylserine exposure.
Sigma-Aldrich
Gly-Pro-Arg-Pro amide, ≥97% (HPLC)