Merck
  • Upregulation of OASIS/CREB3L1 in podocytes contributes to the disturbance of kidney homeostasis.
Upregulation of OASIS/CREB3L1 in podocytes contributes to the disturbance of kidney homeostasis.
Communications biology (2022-07-23)
Yoshiaki Miyake, Masanori Obana, Ayaha Yamamoto, Shunsuke Noda, Koki Tanaka, Hibiki Sakai, Narihito Tatsumoto, Chihiro Makino, Soshi Kanemoto, Go Shioi, Shota Tanaka, Makiko Maeda, Yoshiaki Okada, Kazunori Imaizumi, Katsuhiko Asanuma, Yasushi Fujio
ABSTRACT
Podocyte injury is involved in the onset and progression of various kidney diseases. We previously demonstrated that the transcription factor, old astrocyte specifically induced substance (OASIS) in myofibroblasts, contributes to kidney fibrosis, as a novel role of OASIS in the kidneys. Importantly, we found that OASIS is also expressed in podocytes; however, the pathophysiological significance of OASIS in podocytes remains unknown. Upon lipopolysaccharide (LPS) treatment, there is an increase in OASIS in murine podocytes. Enhanced serum creatinine levels and tubular injury, but not albuminuria and podocyte injury, are attenuated upon podocyte-restricted OASIS knockout in LPS-treated mice, as well as diabetic mice. The protective effects of podocyte-specific OASIS deficiency on tubular injury are mediated by protein kinase C iota (PRKCI/PKCι), which is negatively regulated by OASIS in podocytes. Furthermore, podocyte-restricted OASIS transgenic mice show tubular injury and tubulointerstitial fibrosis, with severe albuminuria and podocyte degeneration. Finally, there is an increase in OASIS-positive podocytes in the glomeruli of patients with minimal change nephrotic syndrome and diabetic nephropathy. Taken together, OASIS in podocytes contributes to podocyte and/or tubular injury, in part through decreased PRKCI. The induction of OASIS in podocytes is a critical event for the disturbance of kidney homeostasis.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-OASIS/CREB3L1 Antibody, clone 44C7, clone 44C7, from mouse