Merck
  • An acquired phosphatidylinositol 4-phosphate transport initiates T-cell deterioration and leukemogenesis.
An acquired phosphatidylinositol 4-phosphate transport initiates T-cell deterioration and leukemogenesis.
Nature communications (2022-07-30)
Wenbin Zhong, Weize Lin, Yingjie Yang, Dan Chen, Xiuye Cao, Mengyang Xu, Guoping Pan, Huanzhao Chen, Jie Zheng, Xiaoqin Feng, Li Hua Yang, Chaofeng Lai, Vesa M Olkkonen, Jun Xu, Shuzhong Cui, Daoguang Yan
ABSTRACT
Lipid remodeling is crucial for malignant cell transformation and tumorigenesis, but the precise molecular processes involved and direct evidences for these in vivo remain elusive. Here, we report that oxysterol-binding protein (OSBP)-related protein 4 L (ORP4L) is expressed in adult T-cell leukemia (ATL) cells but not normal T-cells. In ORP4L knock-in T-cells, ORP4L dimerizes with OSBP to control the shuttling of OSBP between the Golgi apparatus and the plasma membrane (PM) as an exchanger of phosphatidylinositol 4-phosphate [PI(4)P]/cholesterol. The PI(4)P arriving at the PM via this transport machinery replenishes phosphatidylinositol 4,5-bisphosphate [PI(4,5)P2] and phosphatidylinositol (3,4,5) trisphosphate [PI(3,4,5)P3] biosynthesis, thus contributing to PI3K/AKT hyperactivation and T-cell deterioration in vitro and in vivo. Disruption of ORP4L and OSBP dimerization disables PI(4)P transport and T-cell leukemogenesis. In summary, we identify a non-vesicular lipid transport machinery between Golgi and PM maintaining the oncogenic signaling competence initiating T-cell deterioration and leukemogenesis.
Sigma-Aldrich
Anti-OSBP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial