Merck
  • Evaluation of the novel antiepileptic drug, AWD 131-138, for benzodiazepine-like discriminative stimulus and reinforcing effects in squirrel monkeys.
Evaluation of the novel antiepileptic drug, AWD 131-138, for benzodiazepine-like discriminative stimulus and reinforcing effects in squirrel monkeys.
European journal of pharmacology (2003-04-19)
Sevil Yasar, Jack Bergman, Patrik Munzar, Godfrey Redhi, Christine Tober, Norbert Knebel, Michael Zschiesche, Carol Paronis
ABSTRACT
AWD 131-138 [1-(4-chlorophenyl)-4-morpholino-imidazolin-2-one], a new low-affinity partial benzodiazepine receptor agonist with potent anticonvulsant and anxiolytic properties in rodent models, was studied in squirrel monkeys trained to discriminate intramuscular (i.m.) injections of midazolam (0.3 mg/kg) from injections of vehicle. Diazepam produced midazolam-like responding at cumulative doses of 1.0 and 3.0 mg/kg i.m. and decreased rates of responding at 3.0 mg/kg (plasma levels of about 400 ng/ml). In contrast, AWD 131-138 did not produce midazolam-like responding or alter response rates at cumulative doses up to 18.0 mg/kg i.m. (plasma levels over 2100 ng/ml). Other monkeys were trained to intravenously (i.v.) self-administer cocaine (56.0 microg/kg/injection). When AWD 131-138 (10-100 microg/kg/injection) was studied by substitution, responding declined to vehicle substitution levels within three sessions. At the dose of 100 microg/kg i.v. AWD 131-138, sufficient drug was self-administered during the first session (about 3.5 mg/kg) to produce plasma levels above 1000 ng/ml, yet responding over the next two sessions dropped to vehicle levels. The failure of AWD 131-138 to produce benzodiazepine-like discriminative effects and the absence of drug self-administration behavior when substituted for cocaine suggest that its abuse liability is low.
Sigma-Aldrich
Imepitoin, ≥98% (HPLC)