Merck
  • Antibody Light Chains: Key to Increased Monoclonal Antibody Yields in Expi293 Cells?
Antibody Light Chains: Key to Increased Monoclonal Antibody Yields in Expi293 Cells?
Antibodies (Basel, Switzerland) (2022-06-02)
Siqi Gong, Seijal Gautam, Joshua D Coneglio, Hanna B Scinto, Ruth M Ruprecht
ABSTRACT
When constructing isogenic recombinant IgM-IgG pairs, we discovered that μ heavy chains strongly prefer partnering with λ light chains for optimal IgM expression in transiently cotransfected Expi293 cells. When μ chains were paired with κ light chains, IgM yields were low but increased by logs-up to 20,000 X-by using λ chains instead. Switching light chains did not alter epitope specificity. For dimeric IgA2, optimal expression involved pairing with λ chains, whereas light-chain preference varied for other immunoglobulin classes. In summary, recombinant IgM production can be drastically increased by using λ chains, an important finding in the use of IgM for mucosal immunoprophylaxis.
Sigma-Aldrich
Goat Anti-Human Ig λ chain Antibody, HRP conjugate, Chemicon®, from goat
Sigma-Aldrich
IgM from human serum, reagent grade, ~95% (HPLC), buffered aqueous solution