Merck
  • Mechanism of Microbial Metabolite Leupeptin in the Treatment of COVID-19 by Traditional Chinese Medicine Herbs.
Mechanism of Microbial Metabolite Leupeptin in the Treatment of COVID-19 by Traditional Chinese Medicine Herbs.
mBio (2021-09-29)
Lifeng Fu, Shuai Shao, Yong Feng, Fei Ye, Xue Sun, Qingling Wang, Feng Yu, Qisheng Wang, Baoying Huang, Peihua Niu, Xuebing Li, Catherine C L Wong, Jianxun Qi, Wenjie Tan, George Fu Gao
ABSTRACT
Coronavirus disease 2019 (COVID-19) has caused huge deaths and economic losses worldwide in the current pandemic. The main protease (Mpro) of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) is thought to be an ideal drug target for treating COVID-19. Leupeptin, a broad-spectrum covalent inhibitor of serine, cysteine, and threonine proteases, showed inhibitory activity against Mpro, with a 50% inhibitory concentration (IC50) value of 127.2 μM in vitro in our study here. In addition, leupeptin can also inhibit SARS-CoV-2 in Vero cells, with 50% effective concentration (EC50) values of 42.34 μM. More importantly, various strains of streptomyces that have a broad symbiotic relationship with medicinal plants can produce leupeptin and leupeptin analogs to regulate autogenous proteases. Fingerprinting and structure elucidation using high-performance liquid chromatography (HPLC) and high-resolution mass spectrometry (HRMS), respectively, further proved that the Qing-Fei-Pai-Du (QFPD) decoction, a traditional Chinese medicine (TCM) formula for the effective treatment of COVID-19 during the period of the Wuhan outbreak, contains leupeptin. All these results indicate that leupeptin at least contributes to the antiviral activity of the QFPD decoction against SARS-CoV-2. This also reminds us to pay attention to the microbiomes in TCM herbs as streptomyces in the soil might produce leupeptin that will later infiltrate the medicinal plant. We propose that plants, microbiome, and microbial metabolites form an ecosystem for the effective components of TCM herbs. IMPORTANCE A TCM formula has played an important role in the treatment of COVID-19 in China. However, the mechanism of TCM action is still unclear. In this study, we identified leupeptin, a metabolite produced by plant-symbiotic actinomyces (PSA), which showed antiviral activity in both cell culture and enzyme assays. Moreover, leupeptin found in the QFPD decoction was confirmed by both HPLC fingerprinting and HRMS. These results suggest that leupeptin likely contributes to the antiviral activity of the QFPD decoction against SARS-CoV-2. This result gives us important insight into further studies of the PSA metabolite and medicinal plant ecosystem for future TCM modernization research.
Sigma-Aldrich
Leupeptin, BioUltra, microbial, ≥95% (HPLC)