Merck
  • Fatty acid oxidation fuels glioblastoma radioresistance with CD47-mediated immune evasion.
Fatty acid oxidation fuels glioblastoma radioresistance with CD47-mediated immune evasion.
Nature communications (2022-03-23)
Nian Jiang, Bowen Xie, Wenwu Xiao, Ming Fan, Shanxiu Xu, Yixin Duan, Yamah Hamsafar, Angela C Evans, Jie Huang, Weibing Zhou, Xuelei Lin, Ningrong Ye, Siyi Wanggou, Wen Chen, Di Jing, Ruben C Fragoso, Brittany N Dugger, Paul F Wilson, Matthew A Coleman, Shuli Xia, Xuejun Li, Lun-Quan Sun, Arta M Monjazeb, Aijun Wang, William J Murphy, Hsing-Jien Kung, Kit S Lam, Hong-Wu Chen, Jian Jian Li
ABSTRACT
Glioblastoma multiforme (GBM) remains the top challenge to radiotherapy with only 25% one-year survival after diagnosis. Here, we reveal that co-enhancement of mitochondrial fatty acid oxidation (FAO) enzymes (CPT1A, CPT2 and ACAD9) and immune checkpoint CD47 is dominant in recurrent GBM patients with poor prognosis. A glycolysis-to-FAO metabolic rewiring is associated with CD47 anti-phagocytosis in radioresistant GBM cells and regrown GBM after radiation in syngeneic mice. Inhibition of FAO by CPT1 inhibitor etomoxir or CRISPR-generated CPT1A-/-, CPT2-/-, ACAD9-/- cells demonstrate that FAO-derived acetyl-CoA upregulates CD47 transcription via NF-κB/RelA acetylation. Blocking FAO impairs tumor growth and reduces CD47 anti-phagocytosis. Etomoxir combined with anti-CD47 antibody synergizes radiation control of regrown tumors with boosted macrophage phagocytosis. These results demonstrate that enhanced fat acid metabolism promotes aggressive growth of GBM with CD47-mediated immune evasion. The FAO-CD47 axis may be targeted to improve GBM control by eliminating the radioresistant phagocytosis-proofing tumor cells in GBM radioimmunotherapy.
Sigma-Aldrich
(+)-Etomoxir sodium salt hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
A-485, ≥98% (HPLC)
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution