Merck
  • CRISPR-Cas9-mediated loss of function of β-catenin attenuates intervertebral disc degeneration.
CRISPR-Cas9-mediated loss of function of β-catenin attenuates intervertebral disc degeneration.
Molecular therapy. Nucleic acids (2022-05-05)
Yunshan Fan, Lan Zhao, Yumei Lai, Ke Lu, Jian Huang
ABSTRACT
Intervertebral disc degeneration is a very common medical condition causing pain and disability, and it cannot be reversed by available treatment options. Here we report that targeting β-catenin, a pivotal factor associated with disc degeneration, ameliorates disc degeneration in a mouse model of disc injury. Degenerative changes in the disc in response to disc injury include decompression of nucleus pulposus (NP), replacement of notochordal cells in the NP by chondrocyte-like cells, and disorganization of annulus fibrosus (AF). Importantly, downregulation of β-catenin through intradiscal injection of CRISPR-Cas9-expressing adeno-associated virus significantly mitigated all these pathological changes, by preserving notochordal cells and attenuating chondro-osteogenesis in the NP, as well as maintaining the AF structure. Moreover, β-catenin loss-of-function decelerated the rapid induction of catabolic reactions in disc matrix and attenuated pain-related neural events during disc degeneration. Thus, our data demonstrate that targeting β-catenin in disc cells through CRISPR-Cas9 has multifaceted therapeutic effects on disc degeneration, and we suggest that β-catenin plays a fundamental role in the remodeling and degenerative processes of the disc. In addition, this study proposes that CRISPR-Cas9 is a useful tool for identifying new drug targets and developing therapeutic strategies for disc degeneration.
Sigma-Aldrich
Anti-Collagen Type I Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid