Merck
  • Concentration dependent effect of GsMTx4 on mechanosensitive channels of small conductance in E. coli spheroplasts.
Concentration dependent effect of GsMTx4 on mechanosensitive channels of small conductance in E. coli spheroplasts.
European biophysics journal : EBJ (2009-01-10)
Annette C Hurst, Philip A Gottlieb, Boris Martinac
ABSTRACT
The spider peptide GsMTx4, at saturating concentration of 5 muM, is an effective and specific inhibitor for stretch-activated mechanosensitive (MS) channels found in a variety of eukaryotic cells. Although the structure of the peptide has been solved, the mode of action remains to be determined. Because of its amphipathic structure, the peptide is proposed to interact with lipids at the boundaries of the MS channel proteins. In addition, GsMTx4 has antimicrobial effects, inhibiting growth of several species of bacteria in the range of 5-64 microM. Previous studies on prokaryotic MS channels, which serve as model systems to explore the principle of MS channel gating, have shown that various amphipathic compounds acting at the protein-lipid interface affect MS channel gating. We have therefore analyzed the effect of different concentrations of extracellular GsMTx4 on MS channels of small conductance, MscS and MscK, in the cytoplasmic membrane of wild-type E. coli spheroplasts using the patch-clamp technique. Our study shows that the peptide GsMTx4 exhibits a biphasic response in which peptide concentration determines inhibition or potentiation of activity in prokaryotic MS channels. At low peptide concentrations of 2 and 4 microM the gating of the prokaryotic MS channels was hampered, manifested by a decrease in pressure sensitivity. In contrast, application of peptide at concentrations of 12 and 20 microM facilitated prokaryotic MS channel opening by increasing the pressure sensitivity.
Sigma-Aldrich
GsMTx4 trifluoroacetate, ≥85% (HPLC)