Merck
  • Single-cell CUT&Tag analysis of chromatin modifications in differentiation and tumor progression.
Single-cell CUT&Tag analysis of chromatin modifications in differentiation and tumor progression.
Nature biotechnology (2021-04-14)
Steven J Wu, Scott N Furlan, Anca B Mihalas, Hatice S Kaya-Okur, Abdullah H Feroze, Samuel N Emerson, Ye Zheng, Kalee Carson, Patrick J Cimino, C Dirk Keene, Jay F Sarthy, Raphael Gottardo, Kami Ahmad, Steven Henikoff, Anoop P Patel
ABSTRACT
Methods for quantifying gene expression1 and chromatin accessibility2 in single cells are well established, but single-cell analysis of chromatin regions with specific histone modifications has been technically challenging. In this study, we adapted the CUT&Tag method3 to scalable nanowell and droplet-based single-cell platforms to profile chromatin landscapes in single cells (scCUT&Tag) from complex tissues and during the differentiation of human embryonic stem cells. We focused on profiling polycomb group (PcG) silenced regions marked by histone H3 Lys27 trimethylation (H3K27me3) in single cells as an orthogonal approach to chromatin accessibility for identifying cell states. We show that scCUT&Tag profiling of H3K27me3 distinguishes cell types in human blood and allows the generation of cell-type-specific PcG landscapes from heterogeneous tissues. Furthermore, we used scCUT&Tag to profile H3K27me3 in a patient with a brain tumor before and after treatment, identifying cell types in the tumor microenvironment and heterogeneity in PcG activity in the primary sample and after treatment.
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys27) Antibody, clone RM172, clone RM172, from rabbit
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit