Merck
  • Determination of 49 organophosphorus pesticide residues and their metabolites in fish, egg, and milk by dual gas chromatography-dual pulse flame photometric detection with gel permeation chromatography cleanup.
Determination of 49 organophosphorus pesticide residues and their metabolites in fish, egg, and milk by dual gas chromatography-dual pulse flame photometric detection with gel permeation chromatography cleanup.
Journal of agricultural and food chemistry (2012-02-04)
Lixin Yang, Heli Li, Fangang Zeng, Yinping Liu, Ruifeng Li, Huijing Chen, Yunfeng Zhao, Hong Miao, Yongning Wu
ABSTRACT
A new method for the quantitative determination of 49 kinds of organophosphorus pesticide residues and their metabolites in fish, egg, and milk by dual gas chromatography-dual pulse flame photometric detection was developed. Homogenized samples were extracted with acetone and methylene chloride (1 + 1, v/v), and then the extracts were cleaned up by gel permeation chromatography (GPC). The response of each organophosphorus pesticide showed a good linearity with its concentration; the linearity correlation was not less than 0.99. The detection limits (S/N = 3) of pesticides were in the range of 0.001-0.025 mg kg⁻¹. The recovery experiments were performed by blank sample spiked at low, medium, and high fortification levels. The recoveries for fish, egg, and milk were 50.9-142.2, 53.3-137.2, and 50.3-139.4% with relative standard deviations (RSD, n = 6) of 2.3-24.9, 4.3-26.7, and 2.8-32.2%, respectively. The method was applied to detect organophosphorus pesticides in samples collected from the market, and satisfactory results were obtained. This quantitative method was highly sensitive and exact and could be applied to the accurate determination of organophosphorus contaminants in fish, egg, and milk.
Product Description
Supelco
Disulfoton-sulfone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland