Merck
  • Development of direct competitive ELISA for residue analysis of fungicide chlorothalonil in vegetables.
Development of direct competitive ELISA for residue analysis of fungicide chlorothalonil in vegetables.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan (2014-07-06)
Fumiko Okazaki, Yuki Hirakawa, Yukie Yamaguchi-Murakami, Ayako Harada, Eiki Watanabe, Seiji Iwasa, Hiroshi Narita, Shiro Miyake
ABSTRACT
A direct competitive (dc)-ELISA was developed for rapid and simple determination of chlorothalonil residue in vegetables. A carboxylic acid derivative of pentachlorophenol was used to prepare an anti-chlorothalonil monoclonal antibody (MoAb) that showed adequate reactivity for dc-ELISA. Before homogenization of vegetable samples, phosphoric acid was added (vegetable-10% phosphoric acid (2 : 1, w/v)) to block enzymatic decomposition of chlorothalonil. The use of phosphate buffer (100 mmol/L, pH 7.0) minimized the influence of phosphoric acid on competitive reaction in the dc-ELISA. Working range was 0.10 to 6.0 ng/mL in the optimized dc-ELISA. The recovery of chlorothalonil spiked in cucumber and eggplant was 97.1 to 125%. The results correlated well with those obtained by HPLC analysis. The dc-ELISA could rapidly determine chlorothalonil after a simple sample preparation procedure.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Albumin from rabbit serum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)