Merck
  • DHODH inhibition modulates glucose metabolism and circulating GDF15, and improves metabolic balance.
DHODH inhibition modulates glucose metabolism and circulating GDF15, and improves metabolic balance.
iScience (2021-06-12)
Juan Zhang, Graciela Terán, Mihaela Popa, Harsha Madapura, Marcus James Graeme Watson Ladds, Danai Lianoudaki, Jacob Grünler, Marie Arsenian-Henriksson, Emmet McCormack, Martin Enrique Rottenberg, Sergiu-Bogdan Catrina, Sonia Laín, Suhas Darekar
ABSTRACT
Dihydroorotate dehydrogenase (DHODH) is essential for the de novo synthesis of pyrimidine ribonucleotides, and as such, its inhibitors have been long used to treat autoimmune diseases and are in clinical trials for cancer and viral infections. Interestingly, DHODH is located in the inner mitochondrial membrane and contributes to provide ubiquinol to the respiratory chain. Thus, DHODH provides the link between nucleotide metabolism and mitochondrial function. Here we show that pharmacological inhibition of DHODH reduces mitochondrial respiration, promotes glycolysis, and enhances GLUT4 translocation to the cytoplasmic membrane and that by activating tumor suppressor p53, increases the expression of GDF15, a cytokine that reduces appetite and prolongs lifespan. In addition, similar to the antidiabetic drug metformin, we observed that in db/db mice, DHODH inhibitors elevate levels of circulating GDF15 and reduce food intake. Further analysis using this model for obesity-induced diabetes revealed that DHODH inhibitors delay pancreatic β cell death and improve metabolic balance.
Sigma-Aldrich
Brequinar sodium salt hydrate, ≥97% (HPLC)
Sigma-Aldrich
Monoclonal Anti-GDF15 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0328, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Anti-MDM2 (Ab-1) Mouse mAb (IF2), liquid, clone IF2, Calbiochem®