Merck
  • RSV infection-elicited high MMP-12-producing macrophages exacerbate allergic airway inflammation with neutrophil infiltration.
RSV infection-elicited high MMP-12-producing macrophages exacerbate allergic airway inflammation with neutrophil infiltration.
iScience (2021-10-28)
Airi Makino, Takehiko Shibata, Mashiro Nagayasu, Ikuo Hosoya, Toshiyo Nishimura, Chihiro Nakano, Kisaburo Nagata, Toshihiro Ito, Yoshimasa Takahashi, Shigeki Nakamura
ABSTRACT
Respiratory syncytial virus (RSV) infection often exacerbates bronchial asthma, but there is no licensed RSV vaccine or specific treatments. Here we show that RSV-induced alveolar macrophages, which produce high levels of matrix metalloproteinase-12 (MMP-12), exacerbate allergic airway inflammation with increased neutrophil infiltration. When mice subjected to allergic airway inflammation via exposure to the house dust mite antigen (HDM) were infected with RSV (HDM/RSV), MMP-12 expression, viral load, neutrophil infiltration, and airway hyperresponsiveness (AHR) were increased compared to those in the HDM and RSV groups. These exacerbations in the HDM/RSV group were attenuated in MMP-12-deficient mice and mice treated with MMP408, a selective MMP-12 inhibitor, but not in mice treated with dexamethasone. Finally, M2-like macrophages produced MMP-12, and its production was promoted by increase of IFN-β-induced IL-4 receptor expression with RSV infection. Thus, targeting MMP-12 represents a potentially novel therapeutic strategy for the exacerbation of asthma.
Sigma-Aldrich
Dexamethasone-Water Soluble, suitable for cell culture, BioReagent
Sigma-Aldrich
MMP-12 Inhibitor, MMP408, The MMP-12 Inhibitor, MMP408, also referenced under CAS 1258003-93-8, controls the biological activity of MMP-12. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.