Merck
  • FRAP: A Powerful Method to Evaluate Membrane Fluidity in Caenorhabditis elegans.
FRAP: A Powerful Method to Evaluate Membrane Fluidity in Caenorhabditis elegans.
Bio-protocol (2018-07-05)
Ranjan Devkota, Marc Pilon
ABSTRACT
FRAP (Fluorescence Recovery After Photobleaching) is probably the most direct method to investigate the dynamics of molecules in living cells. Here, we describe FRAP to quantify membrane fluidity in C. elegans. Using FRAP, we have shown that cold, glucose and exogenous saturated fatty acids can decrease the fluidity of cellular membranes in certain mutants.
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, low EEO
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
α-D-Glucose, anhydrous, 96%
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
(−)-Tetramisole hydrochloride, ≥99% (GC)