Merck
  • Suppression of lysosomal acid alpha-glucosidase impacts the modulation of transcription factor EB translocation in pancreatic cancer.
Suppression of lysosomal acid alpha-glucosidase impacts the modulation of transcription factor EB translocation in pancreatic cancer.
Cancer science (2021-05-02)
Ryoga Hamura, Yoshihiro Shirai, Yohta Shimada, Nobuhiro Saito, Tomohiko Taniai, Takashi Horiuchi, Naoki Takada, Yumi Kanegae, Toru Ikegami, Toya Ohashi, Katsuhiko Yanaga
ABSTRACT
Lysosomal degradation plays a crucial role in the metabolism of biological macromolecules supplied by autophagy. The regulation of the autophagy-lysosome system, which contributes to intracellular homeostasis, chemoresistance, and tumor progression, has recently been revealed as a promising therapeutic approach for pancreatic cancer (PC). However, the details of lysosomal catabolic function in PC cells have not been fully elucidated. In this study, we show evidence that suppression of acid alpha-glucosidase (GAA), one of the lysosomal enzymes, improves chemosensitivity and exerts apoptotic effects on PC cells through the disturbance of expression of the transcription factor EB. The levels of lysosomal enzyme were elevated by gemcitabine in PC cells. In particular, the levels of GAA were responsive to gemcitabine in a dose-dependent and time-dependent manner. Small interfering RNA against the GAA gene (siGAA) suppressed cell proliferation and promoted apoptosis in gemcitabine-treated PC cells. In untreated PC cells, we observed accumulation of depolarized mitochondria. Gene therapy using adenoviral vectors carrying shRNA against the GAA gene increased the number of apoptotic cells and decreased the tumor growth in xenograft model mice. These results indicate that GAA is one of the key targets to improve the efficacy of gemcitabine and develop novel therapies for PC.
Sigma-Aldrich
4-Methylumbelliferyl α-D-glucopyranoside, α-glucosidase substrate
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)