Merck
  • A phase I study of AT-101 with cisplatin and etoposide in patients with advanced solid tumors with an expanded cohort in extensive-stage small cell lung cancer.
A phase I study of AT-101 with cisplatin and etoposide in patients with advanced solid tumors with an expanded cohort in extensive-stage small cell lung cancer.
Investigational new drugs (2013-07-19)
William R Schelman, Tabraiz A Mohammed, Anne M Traynor, Jill M Kolesar, Rebecca M Marnocha, Jens Eickhoff, Michael Keppen, Dona B Alberti, George Wilding, Naoko Takebe, Glenn Liu
ABSTRACT
A phase I, dose-escalation study of AT-101 with cisplatin and etoposide was conducted to determine the maximum tolerated dose (MTD)/recommended phase 2 dose (RP2D), safety and pharmacokinetics in patients with advanced solid tumors, with an expanded cohort in patients with extensive-stage small cell lung cancer (ES-SCLC) to assess preliminary activity. In the dose escalation portion, increasing doses of AT-101 were administered orally BID on days 1-3 along with cisplatin on day 1 and etoposide on days 1-3 of a 21 day cycle. At the RP2D, an additional 7 patients with untreated ES-SCLC were enrolled. Twenty patients were enrolled in the dose-escalation cohort, and 7 patients with ES-SCLC were enrolled in the expanded cohort. The MTD/RP2D was established at AT-101 40 mg BID days 1-3 with cisplatin 60 mg/m2 and etoposide 120 mg/m2 on day 1 of a 21 day cycle with pegfilgrastim support. Two DLTs of neutropenic fever were seen at dose level 1. After the addition of pegfilgrastim, no additional DLTs were observed. Grade 3/4 treatment-related toxicities included: diarrhea, increased AST, neutropenia, hypophosphatemia, hyponatremia, myocardial infarction and pulmonary embolism. No apparent PK interactions were observed between the agents. Preliminary activity was observed with PRs in patients with ES-SCLC, high-grade neuroendocrine tumor, esophageal cancer and NSCLC. AT-101 with cisplatin and etoposide is well tolerated with growth factor support. Anti-tumor activity was observed in a variety of cancers including ES-SCLC, supporting further investigation with BH-3 mimetics in combination with standard chemotherapy for ES-SCLC.
Sigma-Aldrich
(R)-(−)-2-Amino-1-propanol, 98%