Merck
  • Immunomodulatory Activity of Human Bone Marrow and Adipose-Derived Mesenchymal Stem Cells Prolongs Allogenic Skin Graft Survival in Nonhuman Primates.
Immunomodulatory Activity of Human Bone Marrow and Adipose-Derived Mesenchymal Stem Cells Prolongs Allogenic Skin Graft Survival in Nonhuman Primates.
Cell journal (2021-03-03)
Fattah Sotoodehnejadnematalahi, Reza Moghadasali, Mostafa Hajinasrollah, Ehsan Ehsani, Ensiyeh Hajizadeh-Saffar, Niloofar Sodeifi, Reza Saidi, Morteza Zarrabi, Mohammad Farzanehkhah, Bahareh Sadeghi, Hossein Baharvand, Nasser Aghdami
ABSTRACT
In the present study, we examined the tolerance-inducing effects of human adipose-derived mesenchymal stem cells (hAD-MSCs) and bone marrow-derived MSCs (hBM-MSCs) on a nonhuman primate model of skin transplantation. In this experimental study, allogenic and xenogeneic of immunomodulatory properties of human AD-MSCs and BM-MSCs were evaluated by mixed lymphocyte reaction (MLR) assays. Human MSCs were obtained from BM or AD tissues (from individuals of either sex with an age range of 35 to 65 years) and intravenously injected (2×106 MSCs/kg) after allogeneic skin grafting in a nonhuman primate model. The skin sections were evaluated by H and E staining for histopathological evaluations, particularly inflammation and rejection reaction of grafts after 96 hours of cell injection. At the mRNA and protein levels, cellular mediators of inflammation, such as CD4+IL-17+ (T helper 17; Th17) and CD4+INF-γ+ (T helper 1, Th1) cells, along with CD4+FoxP3+ cells (Treg), as the mediators of immunomodulation, were measured by RT-PCR and flow cytometry analyses. A significant Treg cells expansion was observed in MSCs-treated animals which reached the zenith at 24 hours and remained at a high concentration for 96 hours; however, Th1 and Th17 cells were significantly decreased. Our results showed that human MSCs significantly decrease Th1 and Th17 cell proliferation by decreasing interleukin-17 (IL-17) and interferon-γ (INF-γ) production and significantly increase Treg cell proliferation by increasing FoxP3 production. They also extend the allogenic skin graft survival in nonhuman primates. Histological evaluations showed no obvious presence of inflammatory cells or skin redness or even bulging after MSCs injection up to 96 hours, compared to the group without MSCs. There were no significant differences between hBM-MSCs and hAD-MSCs in terms of histopathological scores and inflammatory responses (P<0.05). It seems that MSCs could be regarded as a valuable immunomodulatory tool to reduce the use of immunosuppressive agents.
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Mouse IgG2b Negative Control, clone GC198, FITC conjugate, Mouse IgG2b Negative Control Monoclonal Antibody validated for use in Flow Cytometry & Immunofluorescence.
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt ≥300,000