Merck
  • Metabolically controlled histone H4K5 acylation/acetylation ratio drives BRD4 genomic distribution.
Metabolically controlled histone H4K5 acylation/acetylation ratio drives BRD4 genomic distribution.
Cell reports (2021-07-29)
Mengqing Gao, Jin Wang, Sophie Rousseaux, Minjia Tan, Lulu Pan, Lijun Peng, Sisi Wang, Wenqian Xu, Jiayi Ren, Yuanfang Liu, Martin Spinck, Sophie Barral, Tao Wang, Florent Chuffart, Ekaterina Bourova-Flin, Denis Puthier, Sandrine Curtet, Lisa Bargier, Zhongyi Cheng, Heinz Neumann, Jian Li, Yingming Zhao, Jian-Qing Mi, Saadi Khochbin
ABSTRACT
In addition to acetylation, histones are modified by a series of competing longer-chain acylations. Most of these acylation marks are enriched and co-exist with acetylation on active gene regulatory elements. Their seemingly redundant functions hinder our understanding of histone acylations' specific roles. Here, by using an acute lymphoblastic leukemia (ALL) cell model and blasts from individuals with B-precusor ALL (B-ALL), we demonstrate a role of mitochondrial activity in controlling the histone acylation/acetylation ratio, especially at histone H4 lysine 5 (H4K5). An increase in the ratio of non-acetyl acylations (crotonylation or butyrylation) over acetylation on H4K5 weakens bromodomain containing protein 4 (BRD4) bromodomain-dependent chromatin interaction and enhances BRD4 nuclear mobility and availability for binding transcription start site regions of active genes. Our data suggest that the metabolism-driven control of the histone acetylation/longer-chain acylation(s) ratio could be a common mechanism regulating the bromodomain factors' functional genomic distribution.
Sigma-Aldrich
Ranolazine dihydrochloride, ≥98% (HPLC), powder
Sigma-Aldrich
Oligomycin A, ≥99% (HPLC)
Sigma-Aldrich
Triacsin C from Streptomyces sp.
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium octanoate, ≥99% (capillary GC)
Roche
Nuclease S7, Micrococcal nuclease, from Staphylococcus aureus
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture