Merck
  • Porous polylactic acid-silica hybrids: preparation, characterization, and study of mesenchymal stem cell osteogenic differentiation.
Porous polylactic acid-silica hybrids: preparation, characterization, and study of mesenchymal stem cell osteogenic differentiation.
Macromolecular bioscience (2014-10-11)
Christos Pandis, Sara Trujillo, Joana Matos, Sara Madeira, Joaquín Ródenas-Rochina, Sotiria Kripotou, Apostolos Kyritsis, João F Mano, José Luis Gómez Ribelles
ABSTRACT
A novel approach to reinforce polymer porous membranes is presented. In the prepared hybrid materials, the inorganic phase of silica is synthesized in-situ and inside the pores of aminolyzed polylactic acid (PLA) membranes by sol-gel reactions using tetraethylorthosilicate (TEOS) and glycidoxypropyltrimethoxysilane (GPTMS) as precursors. The hybrid materials present a porous structure with a silica layer covering the walls of the pores while GPTMS serves also as coupling agent between the organic and inorganic phase. The adjustment of silica precursors ratio allows the modulation of the thermomechanical properties. Culture of mesenchymal stem cells on these supports in osteogenic medium shows the expression of characteristic osteoblastic markers and the mineralization of the extracellular matrix.
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Sigma-Aldrich
L-Cysteine hydrochloride, anhydrous, from non-animal source, BioReagent, suitable for cell culture, ≥98.0%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, powder, suitable for cell culture
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Alizarin Red S, certified by the Biological Stain Commission
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
β-Glycerol phosphate disodium salt pentahydrate, ≥98.0% (NT)
Sigma-Aldrich
Perchloric acid, ACS reagent, 70%
Sigma-Aldrich
Dexamethasone-Water Soluble, suitable for cell culture, BioReagent