Merck
  • A novel amino acid signaling process governs glucose-6-phosphatase transcription.
A novel amino acid signaling process governs glucose-6-phosphatase transcription.
iScience (2021-07-20)
Sara Fukushima, Hiroki Nishi, Mikako Kumano, Daisuke Yamanaka, Naoyuki Kataoka, Fumihiko Hakuno, Shin-Ichiro Takahashi
ABSTRACT
Emerging evidence has shown that amino acids act as metabolic regulatory signals. Here, we showed that glucose-6-phosphatase (G6Pase) mRNA levels in cultured hepatocyte models were downregulated in an amino-acid-depleted medium. Inversely, stimulation with amino acids increased G6Pase mRNA levels, demonstrating that G6Pase mRNA level is directly controlled by amino acids in a reversible manner. Promoter assay revealed that these amino-acid-mediated changes in G6Pase mRNA levels were attributable to transcriptional regulation, independent of canonical hormone signaling pathways. Metabolomic analysis revealed that amino acid starvation induces a defect in the urea cycle, decreasing ornithine, a major intermediate, and supplementation of ornithine in an amino-acid-depleted medium fully rescued G6Pase mRNA transcription, similar to the effects of amino acid stimulation. This pathway was also independent of established mammalian target of rapamycin complex 1 pathway. Collectively, we present a hypothetical concept of "metabolic regulatory amino acid signal," possibly mediated by ornithine.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Trypsin inhibitor from chicken egg white, Type II-O, Partially purified ovomucoid, containing ovoinhibitor
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), lyophilized powder
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥97% (HPLC), powder
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder