Merck
  • Efficient synthesis and in vitro antitubercular activity of 1,2,3-triazoles as inhibitors of Mycobacterium tuberculosis.
Efficient synthesis and in vitro antitubercular activity of 1,2,3-triazoles as inhibitors of Mycobacterium tuberculosis.
Bioorganic & medicinal chemistry letters (2011-11-09)
Poovan Shanmugavelan, Sangaraiah Nagarajan, Murugan Sathishkumar, Alagusundaram Ponnuswamy, Perumal Yogeeswari, Dharmarajan Sriram
ABSTRACT
Efficient and rapid synthesis of 1,2,3-triazole derivatives has been achieved via Huisgen's 1,3-dipolar cycloaddition between alkyl/arylazides and diethyl/dimethyl acetylenedicarboxylate in excellent yields under solvent-free conditions. The environmentally friendly solvent-free protocol overcomes the limitations associated with the prevailing time-consuming solution phase protocols and affords the triazoles just in 1-3 min. In vitro antitubercular activity of these triazoles was screened against Mycobacterium tuberculosis H(37)Rv strain. Four of the compounds showed MIC in the range of 1.56-3.13 μg/mL proving their potential activity.
Sigma-Aldrich
Acetylenedicarboxylic acid, 95%
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Sigma-Aldrich
Acetylenedicarboxylic acid monopotassium salt, ≥98%