Merck
  • Do Peer Recovery Specialists Improve Outcomes for Individuals with Substance Use Disorder in an Integrative Primary Care Setting? A Program Evaluation.
Do Peer Recovery Specialists Improve Outcomes for Individuals with Substance Use Disorder in an Integrative Primary Care Setting? A Program Evaluation.
Journal of clinical psychology in medical settings (2019-09-15)
Travis A Cos, Archana Bodas LaPollo, Mara Aussendorf, Jason M Williams, Kimberly Malayter, David S Festinger
ABSTRACT
Peer recovery specialists (PRSs) combine their personal experiences with substance use and recovery with clinical skills to support patients in treatment for or recovery from substance use. This paper provides evaluation findings from a SAMHSA-funded program that integrated a PRS team into a primary care clinic to assess the efficacy of PRS support on patients' substance use, healthcare involvement, and criminal justice involvement. PRSs provided a range of services to patients with histories of incarceration and substance use, including facilitating support groups, providing one-on-one individualized support, and navigating services. Data were collected from PRS-supported patients at intake, discharge, and 6 months post-intake. Results revealed reductions in the percentage of patients using substances in the past 30 days, decreased number of days using alcohol, increased engagement in more medical services after program enrollment, increased school enrollment, and increased rates of employment for PRS-supported patients.
Sigma-Aldrich
S-Acetylmercaptosuccinic anhydride, 96%