Merck
  • Enoxacin induces oxidative metabolism and mitigates obesity by regulating adipose tissue miRNA expression.
Enoxacin induces oxidative metabolism and mitigates obesity by regulating adipose tissue miRNA expression.
Science advances (2020-12-04)
Andréa Livia Rocha, Tanes Imamura de Lima, Gerson Profeta de Souza, Renan Oliveira Corrêa, Danilo Lopes Ferrucci, Bruno Rodrigues, Camila Lopes-Ramos, Daniel Nilsson, Thiago Leite Knittel, Pollyana Ribeiro Castro, Mariane Font Fernandes, Flaviano Dos Santos Martins, Raphael Bessa Parmigiani, Leonardo Reis Silveira, Hernandes F Carvalho, Johan Auwerx, Marco Aurélio R Vinolo, Jeremie Boucher, Marcelo A Mori
ABSTRACT
MicroRNAs (miRNAs) have been implicated in oxidative metabolism and brown/beige adipocyte identity. Here, we tested whether widespread changes in miRNA expression promoted by treatment with the small-molecule enoxacin cause browning and prevent obesity. Enoxacin mitigated diet-induced obesity in mice, and this was associated with increased energy expenditure. Consistently, subcutaneous white and brown adipose tissues and skeletal muscle of enoxacin-treated mice had higher levels of markers associated with thermogenesis and oxidative metabolism. These effects were cell autonomous since they were recapitulated in vitro in murine and human cell models. In preadipocytes, enoxacin led to a reduction of miR-34a-5p expression and up-regulation of its target genes (e.g., Fgfr1, Klb, and Sirt1), thus increasing FGF21 signaling and promoting beige adipogenesis. Our data demonstrate that enoxacin counteracts obesity by promoting thermogenic signaling and inducing oxidative metabolism in adipose tissue and skeletal muscle in a mechanism that involves, at least in part, miRNA-mediated regulation.
Product Description
Viscosity Bath Media, White Oil for use 40 to 80 °C