Merck
  • Positive epistasis between disease-causing missense mutations and silent polymorphism with effect on mRNA translation velocity.
Positive epistasis between disease-causing missense mutations and silent polymorphism with effect on mRNA translation velocity.
Proceedings of the National Academy of Sciences of the United States of America (2021-01-21)
Robert Rauscher, Giovana B Bampi, Marta Guevara-Ferrer, Leonardo A Santos, Disha Joshi, David Mark, Lisa J Strug, Johanna M Rommens, Manfred Ballmann, Eric J Sorscher, Kathryn E Oliver, Zoya Ignatova
ABSTRACT
Epistasis refers to the dependence of a mutation on other mutation(s) and the genetic context in general. In the context of human disorders, epistasis complicates the spectrum of disease symptoms and has been proposed as a major contributor to variations in disease outcome. The nonadditive relationship between mutations and the lack of complete understanding of the underlying physiological effects limit our ability to predict phenotypic outcome. Here, we report positive epistasis between intragenic mutations in the cystic fibrosis transmembrane conductance regulator (CFTR)-the gene responsible for cystic fibrosis (CF) pathology. We identified a synonymous single-nucleotide polymorphism (sSNP) that is invariant for the CFTR amino acid sequence but inverts translation speed at the affected codon. This sSNP in cis exhibits positive epistatic effects on some CF disease-causing missense mutations. Individually, both mutations alter CFTR structure and function, yet when combined, they lead to enhanced protein expression and activity. The most robust effect was observed when the sSNP was present in combination with missense mutations that, along with the primary amino acid change, also alter the speed of translation at the affected codon. Functional studies revealed that synergistic alteration in ribosomal velocity is the underlying mechanism; alteration of translation speed likely increases the time window for establishing crucial domain-domain interactions that are otherwise perturbed by each individual mutation.
Product Description
SAFC
Nutrient Mixture F-12 Ham, powder, with L-glutamine and 0.863 mg/L zinc sulfate, without sodium bicarbonate, Coon′s Modification, suitable for cell culture