Merck
  • Synthesis, antioxidant activities and urease inhibition of some new 1,2,4-triazole and 1,3,4-thiadiazole derivatives.
Synthesis, antioxidant activities and urease inhibition of some new 1,2,4-triazole and 1,3,4-thiadiazole derivatives.
European journal of medicinal chemistry (2010-09-11)
Imtiaz Khan, Sajid Ali, Shahid Hameed, Nasim Hasan Rama, Muhammad Tahir Hussain, Abdul Wadood, Reaz Uddin, Zaheer Ul-Haq, Ajmal Khan, Sajjad Ali, M Iqbal Choudhary
ABSTRACT
New series of 4,5-disubstituted-2,4-dihydro-3H-1,2,4-triazole-3-thiones (8a-j) and 2,5-disubstituted-1,3,4-thiadiazoles (9a-h) were synthesized by dehydrative cyclization of hydrazinecarbothioamide derivatives (7a-k) by refluxing in 4N aqueous sodium hydroxide and by overnight stirring with polyphosphoric acid, respectively. The structures of the newly synthesized compounds were characterized by IR, (1)H NMR, (13)C NMR, elemental analysis and mass spectroscopic studies and the synthesized compounds were screened for their antioxidant and urease inhibition activities. N-(2,4-Dimethylphenyl)-5-(4-nitrophenyl)-1,3,4-thiadiazol-2-amine (9h) showed excellent antioxidant activity more than the standard drug whereas 4-(2,4-dimethylphenyl)-5-(3-nitrophenyl)-2,4-dihydro-3H-1,2,4-triazole-3-thione (8d) and 4-(2,3-dimethylphenyl)-5-phenyl-2,4-dihydro-3H-1,2,4-triazole-3-thione (8e) exhibited potent urease inhibitory activities.
Sigma-Aldrich
Propyl gallate, ≥98%, FCC
Sigma-Aldrich
Propyl gallate, powder
Sigma-Aldrich
Propyl gallate, antioxidant, ≥98.0% (HPLC)
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%