Merck
  • Realizing Hydrogen De/Absorption Under Low Temperature for MgH2 by Doping Mn-Based Catalysts.
Realizing Hydrogen De/Absorption Under Low Temperature for MgH2 by Doping Mn-Based Catalysts.
Nanomaterials (Basel, Switzerland) (2020-09-10)
Ze Sun, Liuting Zhang, Nianhua Yan, Jiaguang Zheng, Ting Bian, Zongming Yang, Shichuan Su
ABSTRACT
Magnesium hydride (MgH2) has been considered as a potential material for storing hydrogen, but its practical application is still hindered by the kinetic and thermodynamic obstacles. Herein, Mn-based catalysts (MnCl2 and Mn) are adopted and doped into MgH2 to improve its hydrogen storage performance. The onset dehydrogenation temperatures of MnCl2 and submicron-Mn-doped MgH2 are reduced to 225 °C and 183 °C, while the un-doped MgH2 starts to release hydrogen at 315 °C. Further study reveals that 10 wt% of Mn is the better doping amount and the MgH2 + 10 wt% submicron-Mn composite can quickly release 6.6 wt% hydrogen in 8 min at 300 °C. For hydrogenation, the completely dehydrogenated composite starts to absorb hydrogen even at room temperature and almost 3.0 wt% H2 can be rehydrogenated in 30 min under 3 MPa hydrogen at 100 °C. Additionally, the activation energy of hydrogenation reaction for the modified MgH2 composite significantly decreases to 17.3 ± 0.4 kJ/mol, which is much lower than that of the primitive MgH2. Furthermore, the submicron-Mn-doped sample presents favorable cycling stability in 20 cycles, providing a good reference for designing and constructing efficient solid-state hydrogen storage systems for future application.
Sigma-Aldrich
Oleic acid, technical grade, 90%
SAFC
Oleylamine, ≥98% (primary amine)