Merck
  • Tryptophan, a non-canonical melanin precursor: New L-tryptophan based melanin production by Rubrivivax benzoatilyticus JA2.
Tryptophan, a non-canonical melanin precursor: New L-tryptophan based melanin production by Rubrivivax benzoatilyticus JA2.
Scientific reports (2020-06-04)
Shabbir Ahmad, Mujahid Mohammed, Lakshmi Prasuna Mekala, Sasikala Chintalapati, Venkata Ramana Chintalapati
ABSTRACT
Melanins are chemically diverse ubiquitous pigments found across the life forms synthesized via different biochemical pathways mainly from L-tyrosine or acetyl CoA. Though few reports suggest the possibility of tryptophan-based melanin synthesis, however, such tryptophan-based melanin and its biosynthesis remained a biochemical riddle. Here we report tryptophan-based melanin production by bacterium, Rubrivivax benzoatilyticus JA2. Aerobic cultures of strain JA2 produced brown pigment when grown on L-tryptophan-containing media. Purified pigment showed typical physico-chemical properties of melanin. Further, extensive spectroscopic studies revealed that pigment is an amorphous, indole-type polymer with stable free radical centers. Further, hydrolysis of the brown pigment revealed the presence of indole moiety, confirming the indolic nature of the pigment. Demonstration of in vitro and in vivo pigment synthesis directly from L-tryptophan or hydroxytryptophan confirms tryptophan-based melanin synthesis in strain JA2. Interestingly, canonical melanin biosynthetic inhibitors did not affect the pigment synthesis indicating possible non-canonical tryptophan-based melanin biosynthesis in strain JA2. Further, the exometabolite profiling and precursor feeding studies suggests that L-tryptophan converted to hydroxytryptophan/hydroxyindoles and their subsequent polymerization lead to the formation of melanin. The current study sheds light on biosynthetic diversity of melanins and L-tryptophan can be a potential precursor for melanin synthesis in life forms.
Sigma-Aldrich
5-Hydroxyindole-3-acetic acid, ≥98% (HPLC), crystalline
Supelco
5-Hydroxyindole-3-acetic acid, analytical standard
Sigma-Aldrich
Allyl methyl sulfone, 96%