Merck
  • microRNA-9-5p alleviates blood-brain barrier damage and neuroinflammation after traumatic brain injury.
microRNA-9-5p alleviates blood-brain barrier damage and neuroinflammation after traumatic brain injury.
Journal of neurochemistry (2020-01-18)
Jingchuan Wu, Junchi He, Xiaocui Tian, Yuetao Luo, Jianjun Zhong, Hongrong Zhang, Hui Li, Bo Cen, Tao Jiang, Xiaochuan Sun
ABSTRACT
The level of microRNA-9-5p (miRNA-9-5p) in brain tissues is significantly changed after traumatic brain injury (TBI). However, the effect of miRNA-9-5p for brain function in TBI has not been elucidated. In this study, a controlled cortical impact model was used to induce TBI in Sprague-Dawley rats, and an oxygen glucose deprivation model was used to mimic the pathological state in vitro. Brain microvascular endothelial cells (BMECs) and astrocytes were extracted from immature Sprague-Dawley rats and cocultured to reconstruct blood-brain barrier (BBB) in vitro. The results show that the level of miRNA-9-5p was significantly increased in brain tissues after TBI, and up-regulation of miRNA9-5p contributed to the recovery of neurological function. Up-regulation of miRNA-9-5p with miRNA agomir may significantly alleviate apoptosis, neuroinflammation, and BBB damage in rats after TBI. Moreover, a dual luciferase reporter assay confirmed that miRNA-9-5p is a post-transcriptional modulator of Ptch-1. In in vitro experiments, the results confirmed that up-regulation of miRNA-9-5p with miRNA mimic alleviates cellular apoptosis, inflammatory response, and BBB damage mainly by inhibiting Ptch-1. In addition, we found that the activation of Hedgehog pathway was accompanied by inhibition of NF-κB/MMP-9 pathway in the BMECs treated with miRNA-9-5p mimic. Taken together, these results indicate that up-regulation of miRNA-9-5p alleviates BBB damage and neuroinflammatory responses by activating the Hedgehog pathway and inhibiting NF-κB/MMP-9 pathway, which promotes the recovery of neurological function after TBI.
Sigma-Aldrich
Evans Blue, Dye content ≥75 %
Sigma-Aldrich
Peroxidase from horseradish, Type VI, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
Supelco
Sufentanil Citrate solution, 100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Isoflurane
Sigma-Aldrich
Phosphate Buffered Saline, 10× PBS for Western blots and IP
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%