Merck
  • CCAAT/enhancer binding protein β induces motility and invasion of glioblastoma cells through transcriptional regulation of the calcium binding protein S100A4.
CCAAT/enhancer binding protein β induces motility and invasion of glioblastoma cells through transcriptional regulation of the calcium binding protein S100A4.
Oncotarget (2015-03-05)
Diana Aguilar-Morante, Jose A Morales-Garcia, Angel Santos, Ana Perez-Castillo
ABSTRACT
We have previously shown that decreased expression of CCAAT/enhancer binding protein β (C/EBPβ) inhibits the growth of glioblastoma cells and diminishes their transformation capacity and migration. In agreement with this, we showed that C/EBPβ depletion decreases the mRNA levels of different genes involved in metastasis and invasion. Among these, we found S100 calcium binding protein A4 (S100A4) to be almost undetectable in glioblastoma cells deficient in C/EBPβ. Here, we have evaluated the possible role of S100A4 in the observed effects of C/EBPβ in glioblastoma cells and the mechanism through which S100A4 levels are controlled by C/EBPβ. Our results show that C/EBPβ suppression significantly reduced the levels of S100A4 in murine GL261 and human T98G glioblastoma cells. By employing an S100A4-promoter reporter, we observed a significant induction in the transcriptional activation of the S100A4 gene by C/EBPβ. Furthermore, overexpression of S100A4 in C/EBPβ-depleted glioblastoma cells reverses the enhanced migration and motility induced by this transcription factor. Our data also point to a role of S100A4 in glioblastoma cell invasion and suggest that the C/EBPβ gene controls the invasive potential of GL261 and T98G cells through direct regulation of S100A4. Finally, this study indicates a role of C/EBPβ on the maintenance of the stem cell population present in GL261 glioblastoma cells.
Sigma-Aldrich
Rapid Cell Proliferation Kit