Merck
Novel ABCA1 peptide agonists with antidiabetic action.
Molecular and cellular endocrinology (2018-10-06)
Salman Azhar, Stefanie Bittner, Jie Hu, Wen-Jun Shen, Yuan Cortez, Xiao Hao, Lu Han, Jens O Lagerstedt, Fredric B Kraemer, Jan O Johansson
ABSTRACT
Previously, apoE-derived ABCA1 agonist peptides have been shown to possess anti-atherosclerotic and possibly antidiabetic properties. Here we assessed the in vitro and in vivo actions of a second generation of ABCA1 peptide agonists, CS6253 and T6991-2, on glucose homeostasis. The results show that these two peptides improve glucose tolerance in a prediabetic diet-induced obesity mouse model by enhancing insulin secretion. It was further demonstrated that T6991-2 also improved glucose tolerance in leptin-deficient (ob/ob) mice. CS6253 increased insulin secretion both under basal conditions and in response to high glucose stimulation in pancreatic INS-1 β-cells rendered leptin receptor deficient with specific siRNA. Additional in vitro cell studies suggest that the CS6253 agonist attenuates hepatic gluconeogenesis and glucose transport. It also potentiates insulin-stimulated glucose uptake and utilization. These observed anti-diabetic actions suggest additional benefits of the CS6253 and T6991-2 ABCA1 peptide agonists for cardiovascular disease beyond their direct anti-atherosclerosis properties previously described.
Product Description
Millipore
MILLIPLEX® Mouse Adipokine Magnetic Bead Panel - Endocrine Multiplex Assay, The analytes available for this multiplex kit are: IL-6, Insulin, Leptin, MCP-1, PAI-1 (Total), Resistin, TNF-α.