Merck
  • Trifluoromethyl Thianthrenium Triflate: A Readily Available Trifluoromethylating Reagent with Formal CF3+, CF3•, and CF3- Reactivity.
Trifluoromethyl Thianthrenium Triflate: A Readily Available Trifluoromethylating Reagent with Formal CF3+, CF3•, and CF3- Reactivity.
Journal of the American Chemical Society (2021-05-15)
Hao Jia, Andreas P Häring, Florian Berger, Li Zhang, Tobias Ritter
ABSTRACT
Here we report the synthesis and application of trifluoromethyl thianthrenium triflate (TT-CF3+OTf-) as a novel trifluoromethylating reagent, which is conveniently accessible in a single step from thianthrene and triflic anhydride. We demonstrate the use of TT-CF3+OTf- in electrophilic, radical, and nucleophilic trifluoromethylation reactions.
Sigma-Aldrich
S-(Trifluoromethyl)thianthrenium triflate, ≥95%