Merck
  • Radioprotective effect of chloropyllin, protoporphyrin-IX and bilirubin compared with amifostine® in Drosophila melanogaster.
Radioprotective effect of chloropyllin, protoporphyrin-IX and bilirubin compared with amifostine® in Drosophila melanogaster.
Environmental toxicology and pharmacology (2020-08-05)
E Jiménez, E Pimentel, M P Cruces, A Amaya-Chávez
ABSTRACT
The identification of substances that prevent or minimize the detrimental effects of ionizing radiation is an essential undertaking. The aim of this paper was to evaluate and compare the radioprotective potential of chlorophyllin, protoporphyrin and bilirubin, with amifostine®, an US Food & Drug Administration approved radioprotector Using the somatic mutation and recombination assay in the Drosophila melanogaster wing, it was found that pretreatment (1-9 h) with any of the porphyrins or amifostine® alone, did not affect the larva-adult viability or the basal frequency of mutation. However, they were associated with significant reductions in frequency of somatic mutation and recombination compared with the gamma-irradiated (20 Gy) control as follows: bilirubin (69.3 %)> chlorophyllin (40.0 %)> protoporphyrin (39.0 %)> amifostine® (19.7 %). Bilirubin also caused a 16 % increase in larva-adult viability with 3 h of pretreatment respect to percentage induced in 20 Gy control group. Whilst amifostine® was associated with lower genetic damage after pre-treatment of 1 and 3 h, this did not attain significance. These findings suggest that the tested porphyrins may have some potential as radioprotectant agents.
Product Description
Millipore
Bilirubin, Principal pigment of bile and one of the major end products of hemoglobin decomposition.