Merck
Synthesis of mangiferin, isomangiferin, and homomangiferin.
The Journal of organic chemistry (2010-08-14)
Zhongtao Wu, Guo Wei, Gaoyan Lian, Biao Yu
ABSTRACT
Mangiferin, isomangiferin, and homomangiferin, the xanthone C-glycosides with a wide spectrum of pharmacological effects, were synthesized concisely, featuring a C-glycosylation of a xanthene derivative with perbenzylglucopyranosyl N-phenyltrifluoroacetimidate.
Sigma-Aldrich
Mangiferin Mangifera indica