- Storage media for avulsed teeth: a literature review.
Storage media for avulsed teeth: a literature review.
Dental avulsion is the most severe type of traumatic tooth injuries because it causes damage to several structures and results in the complete displacement of the tooth from its socket in the alveolar bone. The ideal situation is to replant an exarticulated tooth immediately after avulsion because the extraoral time is a determinant factor for treatment success and for a good prognosis. However, it is not always possible. The success of replantation depends on a number of factors that may contribute to accelerate or minimize the occurrence of root resorption or ankylosis, among which is the type and characteristics of the medium used for temporary storage during the time elapsed between avulsion and replantation. Maintaining the tooth in an adequate wet medium that can preserve, as longer as possible, the vitality of the periodontal ligament cells that remain on root surface is the key to success of replantation. Recent research has led to the development of storage media that produce conditions that closely resemble the original socket environment, with adequate osmolality (cell pressure), pH, nutritional metabolites and glucose, and thus create the best possible conditions for storage. Although these storage media can now be purchased in the form of retail products, the most common scenario is that such a product will not be readily available at the moment of the accident This paper reviews the literature on the different storage media that have been investigated for avulsed teeth based on full-length papers retrieved from PubMed/Medline, Lilacs, BBO and SciELO electronic databases using the key words 'storage medium', 'transportation medium', 'avulsion', 'tooth avulsion', 'replantation', 'tooth replantation', 'milk' and 'propolis'. After application of inclusion and exclusion criteria, 39 papers were selected and critically reviewed with respect to the characteristics, efficacy and ease of access of the storage medium. The review of the literature showed that a wide array of types of wet storage media have been evaluated in laboratory studies and clinical reports, including cell and tissue culture solutions like Hank's Balanced Salt Solution (HBSS); medical/hospital products developed specifically for organ storage purposes, such as Viaspan® and Euro-Collins®; culture media, like Minimum Essential Medium (MEM); saline; natural products like water, saliva, bovine milk and its variations, propolis, green tea, Morus rubra (red mulberry), egg white and coconut water; rehydrating solutions, like Gatorade® and Ricetral, and even contact lens solutions. Based on the literature, it could be stated that, so far, apart from Based on the literature, it could be stated that, so far, apart from solutions designed specifically for storage and culture purposes, regular pasteurized whole milk is the most frequently recommended and with the best prognosis among other solutions that are likely to be available at the scene of an accident, such as water, saline or saliva. Its advantages include its high availability, ready accessibility, physiologically compatible pH and osmolality (fluid pressure) with the root-surface adhered PDL cells, presence of nutrients and growth factors. However, there is not yet a single solution that fulfills all requirements to be considered as the ideal medium for temporary storage of avulsed teeth, and research on this field should carry on.