Merck
  • Synthesis of N-Alkyl Anilines from Arenes via Iron-Promoted Aromatic C-H Amination.
Synthesis of N-Alkyl Anilines from Arenes via Iron-Promoted Aromatic C-H Amination.
Organic letters (2021-02-06)
Eric Falk, Valentina C M Gasser, Bill Morandi
ABSTRACT
We report both an intermolecular C-H amination of arenes to access N-methylanilines and an intramolecular variant for the synthesis of tetrahydroquinolines. A newly developed, highly electrophilic aminating reagent was key for the direct synthesis of unprotected N-methylanilines from simple arenes. The reactions display a broad functional group tolerance and employ catalytic amounts of a benign iron salt under mild reaction conditions.