Merck
  • Pseudosterase activity-based specific detection of human serum albumin on gel.
Pseudosterase activity-based specific detection of human serum albumin on gel.
Talanta (2021-01-01)
Deepak Kumar, Rajasri Bhattacharyya, Dibyajyoti Banerjee
ABSTRACT
Human serum albumin (HSA) has pseudoesterase activity. So far on gel specific detection of such property of HSA is never reported. Moreover, protein binding dyes are non-specific for albumin. However, many of such dyes are used for HSA detection. So, dye-based albumin detection on the gel is expected to generate false-positive results for HSA. In this context, we have discovered that Fast Blue BB (FBBB, 0.12%) stains specifically HSA pseudoesterase activity with 2 Naphthyl acetate (2NA) as an ester substrate. Further, neostigmine has not inhibited the pseudoesterase activity associated with HSA. Neostigmine is a known inhibitor of many true esterases like acetylcholinesterase. So, neostigmine addition offers specificity to the method developed for staining of HSA. Additionally, 2NA stains HSA better than bovine serum albumin (BSA). Exploring all these novel findings, we have devised a simple method of HSA detection on the gel, accurately where other esterases are not detected. To the best of our knowledge, our method is the first to detect HSA pseudoesterase activity specifically on gel without getting interfered by any other esterase activity. The method detects HSA better than BSA. We feel that this method will go a long way for the specific detection of HSA on the gel. It is also relevant for understanding the purity of donor human milk matrix and pharmaceutical preparation of HSA. Our method can detect 7 μM of added HSA in human urine. Therefore, our method can be proceeded further for microalbuminuria detection in days to come.
Sigma-Aldrich
Albumin from human serum, lyophilized powder, essentially protease free, ≥96% (agarose gel electrophoresis)